Assay Details
| Gene Symbol: LOC107277011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776333.2 | XP_015631819.1 | 19 - 20 | 3314 | 72 | |
| GenBank mRNA | AK288069.1 | - | 7 - 8 | 954 | 72 | |
More Information
Set Membership: |
|

LOC107277011
Genomic Map