Assay Details
| Gene Symbol: LOC4335760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778631.2 | XP_015634117.1 | 1 - 2 | 316 | 83 | |
| GenBank mRNA | AK071186.1 | - | 1564 | 83 | ||
| AK121434.1 | - | 1 - 2 | 309 | 83 | ||
| AY224583.1 | - | 1 - 2 | 239 | 83 | ||
More Information
Set Membership: |
|

LOC4335760
Genomic Map