Assay Details
| Gene Symbol: LOC9267158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781696.2 | XP_015637182.1 | 4 - 5 | 1571 | 62 | |
| GenBank mRNA | AK287825.1 | - | 3 - 4 | 1254 | 62 | |
| FJ660627.1 | - | 2 - 3 | 147 | 62 | ||
More Information
Set Membership: |
|

LOC9267158
Genomic Map