Assay Details
| Gene Symbol: LOC4339823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783936.2 | XP_015639422.1 | 2 - 3 | 186 | 120 | |
| GenBank mRNA | AK063535.1 | - | 173 | 120 | ||
| AK072845.1 | - | 2 - 3 | 172 | 120 | ||
More Information
Set Membership: |
|

LOC4339823
Genomic Map