Assay Details
| Gene Symbol: LOC4339823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783936.2 | XP_015639422.1 | 9 - 10 | 926 | 63 | |
| GenBank mRNA | AK072845.1 | - | 9 - 10 | 912 | 63 | |
More Information
Set Membership: |
|

LOC4339823
Genomic Map