Assay Details
| Gene Symbol: LOC4339900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788250.1 | XP_015643736.1 | 1 - 2 | 235 | 104 | |
| GenBank mRNA | AK061483.1 | - | 1 - 2 | 133 | 104 | |
| AK074027.1 | - | 1 - 2 | 128 | 104 | ||
| AK099246.1 | - | 1 - 2 | 132 | 104 | ||
| AK104301.1 | - | 1 - 2 | 129 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788250.1 transcripts.
Set Membership: |
|

LOC4339900
Genomic Map