Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 11 - 12 | 1813 | 80 | |
| XM_015794954.2 | XP_015650440.1 | 10 - 11 | 1771 | 80 | ||
| XM_015794955.1 | XP_015650441.1 | 9 - 10 | 1599 | 80 | ||
| GenBank mRNA | AK064832.1 | - | 9 - 10 | 1567 | 80 | |
| AK065120.1 | - | 10 - 11 | 1734 | 80 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map