Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 12 - 13 | 1917 | 65 | |
| XM_015794954.2 | XP_015650440.1 | 11 - 12 | 1875 | 65 | ||
| XM_015794955.1 | XP_015650441.1 | 10 - 11 | 1703 | 65 | ||
| GenBank mRNA | AK064832.1 | - | 10 - 11 | 1671 | 65 | |
| AK065120.1 | - | 11 - 12 | 1838 | 65 | ||
| AY772415.1 | - | 11 - 12 | 1824 | 65 | ||
| AY772416.1 | - | 10 - 11 | 1653 | 65 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map