Assay Details
Gene Symbol: LOC4345945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794953.2 | XP_015650439.1 | 14 - 15 | 2212 | 79 | |
XM_015794954.2 | XP_015650440.1 | 13 - 14 | 2170 | 79 | ||
XM_015794955.1 | XP_015650441.1 | 12 - 13 | 1998 | 79 | ||
GenBank mRNA | AK064832.1 | - | 12 - 13 | 1966 | 79 | |
AK065120.1 | - | 13 - 14 | 2133 | 79 | ||
AY772415.1 | - | 13 - 14 | 2119 | 79 | ||
AY772416.1 | - | 12 - 13 | 1948 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
