Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 3 - 4 | 535 | 67 | |
| XM_015794954.2 | XP_015650440.1 | 3 - 4 | 535 | 67 | ||
| XM_015794955.1 | XP_015650441.1 | 3 - 4 | 534 | 67 | ||
| GenBank mRNA | AK064832.1 | - | 3 - 4 | 502 | 67 | |
| AK065120.1 | - | 3 - 4 | 498 | 67 | ||
| AY772415.1 | - | 3 - 4 | 484 | 67 | ||
| AY772416.1 | - | 3 - 4 | 484 | 67 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map