Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 5 - 6 | 984 | 58 | |
| XM_015794954.2 | XP_015650440.1 | 5 - 6 | 984 | 58 | ||
| XM_015794955.1 | XP_015650441.1 | 5 - 6 | 983 | 58 | ||
| GenBank mRNA | AK064832.1 | - | 5 - 6 | 951 | 58 | |
| AK065120.1 | - | 5 - 6 | 947 | 58 | ||
| AY772415.1 | - | 5 - 6 | 933 | 58 | ||
| AY772416.1 | - | 5 - 6 | 933 | 58 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map