Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794954.2 | XP_015650440.1 | 6 - 7 | 1091 | 90 | |
| GenBank mRNA | AK065120.1 | - | 6 - 7 | 1054 | 90 | |
| AY772415.1 | - | 6 - 7 | 1040 | 90 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map