Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 8 - 9 | 1310 | 65 | |
| XM_015794954.2 | XP_015650440.1 | 7 - 8 | 1268 | 65 | ||
| GenBank mRNA | AK065120.1 | - | 7 - 8 | 1231 | 65 | |
| AY772415.1 | - | 7 - 8 | 1217 | 65 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map