Assay Details
| Gene Symbol: LOC4345945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794953.2 | XP_015650439.1 | 9 - 10 | 1473 | 124 | |
| XM_015794954.2 | XP_015650440.1 | 8 - 9 | 1431 | 124 | ||
| XM_015794955.1 | XP_015650441.1 | 7 - 8 | 1259 | 124 | ||
| GenBank mRNA | AK064832.1 | - | 7 - 8 | 1227 | 124 | |
| AK065120.1 | - | 8 - 9 | 1394 | 124 | ||
| AY772415.1 | - | 8 - 9 | 1380 | 124 | ||
| AY772416.1 | - | 7 - 8 | 1209 | 124 | ||
More Information
Set Membership: |
|

LOC4345945
Genomic Map