Assay Details
| Gene Symbol: LOC4346956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755125.2 | XP_015610611.1 | 4 - 5 | 544 | 119 | |
| XM_015755126.2 | XP_015610612.1 | 4 - 5 | 544 | 119 | ||
| GenBank mRNA | AK069644.1 | - | 4 - 5 | 527 | 119 | |
More Information
Set Membership: |
|

LOC4346956
Genomic Map