Assay Details
| Gene Symbol: LOC4347851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756476.1 | XP_015611962.1 | 1 - 2 | 131 | 61 | |
| GenBank mRNA | AK071381.1 | - | 1 - 2 | 118 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756476.1 transcripts.
Set Membership: |
|

LOC4347851
Genomic Map