Assay Details
| Gene Symbol: LOC4347851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756476.1 | XP_015611962.1 | 2 - 3 | 335 | 76 | |
| XM_015756477.1 | XP_015611963.1 | 2 - 3 | 235 | 76 | ||
| GenBank mRNA | AK071381.1 | - | 2 - 3 | 322 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756477.1 transcripts.
Set Membership: |
|

LOC4347851
Genomic Map