Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 12 - 13 | 1250 | 128 | |
| XM_015761985.2 | XP_015617471.1 | 11 - 12 | 1160 | 128 | ||
| GenBank mRNA | AK101592.1 | - | 11 - 12 | 1155 | 128 | |
More Information
Set Membership: |
|

LOC4349917
Genomic Map