Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 1 - 2 | 150 | 146 | |
| XM_015761985.2 | XP_015617471.1 | 1 - 2 | 150 | 146 | ||
| GenBank mRNA | AK064320.1 | - | 1 - 2 | 126 | 146 | |
| AK101592.1 | - | 1 - 2 | 145 | 146 | ||
More Information
Set Membership: |
|

LOC4349917
Genomic Map