Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 3 - 4 | 409 | 103 | |
| XM_015761985.2 | XP_015617471.1 | 3 - 4 | 409 | 103 | ||
| GenBank mRNA | AK064320.1 | - | 3 - 4 | 385 | 103 | |
| AK101592.1 | - | 3 - 4 | 404 | 103 | ||
More Information
Set Membership: |
|

LOC4349917
Genomic Map