Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 5 - 6 | 559 | 89 | |
| XM_015761985.2 | XP_015617471.1 | 5 - 6 | 559 | 89 | ||
| GenBank mRNA | AK064320.1 | - | 5 - 6 | 535 | 89 | |
| AK101592.1 | - | 5 - 6 | 554 | 89 | ||
More Information
Set Membership: |
|

LOC4349917
Genomic Map