Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 10 - 11 | 1091 | 72 | |
| XM_015761985.2 | XP_015617471.1 | 9 - 10 | 1001 | 72 | ||
| GenBank mRNA | AK064320.1 | - | 10 - 11 | 1067 | 72 | |
| AK101592.1 | - | 9 - 10 | 996 | 72 | ||
More Information
Set Membership: |
|

LOC4349917
Genomic Map