Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 10 - 11 | 1075 | 79 | |
| XM_015771323.2 | XP_015626809.1 | 10 - 11 | 1059 | 79 | ||
| GenBank mRNA | AK100446.1 | - | 10 - 11 | 968 | 79 | |
| D78136.1 | - | 9 - 10 | 925 | 79 | ||
| D85751.1 | - | 10 - 11 | 1023 | 79 | ||
More Information
Set Membership: |
|

LOC4331112
Genomic Map