Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 11 - 12 | 1139 | 93 | |
| XM_015771323.2 | XP_015626809.1 | 11 - 12 | 1123 | 93 | ||
| GenBank mRNA | AK100446.1 | - | 11 - 12 | 1032 | 93 | |
| D78136.1 | - | 10 - 11 | 989 | 93 | ||
| D85751.1 | - | 11 - 12 | 1087 | 93 | ||
More Information
Set Membership: |
|

LOC4331112
Genomic Map