Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 16 - 17 | 1593 | 86 | |
| XM_015771323.2 | XP_015626809.1 | 16 - 17 | 1577 | 86 | ||
| GenBank mRNA | AK058270.1 | - | 5 - 6 | 389 | 86 | |
| AK100446.1 | - | 16 - 17 | 1486 | 86 | ||
| D78136.1 | - | 15 - 16 | 1443 | 86 | ||
| D85751.1 | - | 16 - 17 | 1541 | 86 | ||
More Information
Set Membership: |
|

LOC4331112
Genomic Map