Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 2 - 3 | 332 | 66 | |
| XM_015771323.2 | XP_015626809.1 | 2 - 3 | 316 | 66 | ||
| GenBank mRNA | AK100446.1 | - | 2 - 3 | 225 | 66 | |
| D85751.1 | - | 2 - 3 | 280 | 66 | ||
More Information
Set Membership: |
|

LOC4331112
Genomic Map