Assay Details
| Gene Symbol: LOC4331226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769858.2 | XP_015625344.1 | 1 - 2 | 208 | 111 | |
| XM_015769859.2 | XP_015625345.1 | 1 - 2 | 205 | 111 | ||
| XM_015769860.2 | XP_015625346.1 | 1 - 2 | 206 | 111 | ||
| XM_015769861.2 | XP_015625347.1 | 1 - 2 | 204 | 111 | ||
| XM_015769862.2 | XP_015625348.1 | 1 - 2 | 204 | 111 | ||
| GenBank mRNA | AK099911.1 | - | 1 - 2 | 146 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769858.2, XM_015769859.2, XM_015769860.2, XM_015769861.2, XM_015769862.2 transcripts.
Set Membership: |
|

LOC4331226
Genomic Map