Assay Details
| Gene Symbol: LOC4331226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769858.2 | XP_015625344.1 | 2 - 3 | 324 | 81 | |
| XM_015769859.2 | XP_015625345.1 | 2 - 3 | 321 | 81 | ||
| XM_015769860.2 | XP_015625346.1 | 2 - 3 | 322 | 81 | ||
| XM_015769861.2 | XP_015625347.1 | 2 - 3 | 320 | 81 | ||
| XM_015769862.2 | XP_015625348.1 | 2 - 3 | 320 | 81 | ||
More Information
Set Membership: |
|

LOC4331226
Genomic Map