Assay Details
| Gene Symbol: LOC4331226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769858.2 | XP_015625344.1 | 4 - 5 | 430 | 104 | |
| XM_015769859.2 | XP_015625345.1 | 4 - 5 | 427 | 104 | ||
| XM_015769860.2 | XP_015625346.1 | 4 - 5 | 428 | 104 | ||
| XM_015769861.2 | XP_015625347.1 | 4 - 5 | 426 | 104 | ||
| XM_015769862.2 | XP_015625348.1 | 4 - 5 | 426 | 104 | ||
| GenBank mRNA | AK099911.1 | - | 4 - 5 | 368 | 104 | |
More Information
Set Membership: |
|

LOC4331226
Genomic Map