Assay Details
| Gene Symbol: LOC4331382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773545.2 | XP_015629031.1 | 10 - 11 | 1844 | 67 | |
| XM_015773546.2 | XP_015629032.1 | 10 - 11 | 1844 | 67 | ||
| GenBank mRNA | AK063381.1 | - | 2 - 3 | 408 | 67 | |
More Information
Set Membership: |
|

LOC4331382
Genomic Map