Assay Details
| Gene Symbol: LOC4333323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774581.2 | XP_015630067.1 | 2 - 3 | 446 | 125 | |
| GenBank mRNA | AK070980.1 | - | 2 - 3 | 453 | 125 | |
More Information
Set Membership: |
|

LOC4333323
Genomic Map