Assay Details
| Gene Symbol: LOC4333337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774816.2 | XP_015630302.1 | 12 - 13 | 2268 | 101 | |
| XM_015774817.2 | XP_015630303.1 | 11 - 12 | 2142 | 101 | ||
| GenBank mRNA | AK066278.1 | - | 12 - 13 | 2148 | 101 | |
| AK072334.1 | - | 12 - 13 | 2274 | 101 | ||
| AK121076.1 | - | 1 - 2 | 249 | 101 | ||
| Gene Symbol: LOC4346696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756031.2 | XP_015611517.1 | 11 - 12 | 2159 | 101 | |
More Information
Set Membership: |
|

LOC4333337
Genomic Map