Assay Details
| Gene Symbol: LOC4333337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774816.2 | XP_015630302.1 | 3 - 4 | 909 | 152 | |
| XM_015774817.2 | XP_015630303.1 | 3 - 4 | 909 | 152 | ||
| GenBank mRNA | AK066278.1 | - | 3 - 4 | 789 | 152 | |
| AK072334.1 | - | 3 - 4 | 915 | 152 | ||
| Gene Symbol: LOC4346696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756031.2 | XP_015611517.1 | 3 - 4 | 887 | 152 | |
More Information
Set Membership: |
|

LOC4333337
Genomic Map