Assay Details
| Gene Symbol: LOC4334435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772846.2 | XP_015628332.1 | 11 - 12 | 4252 | 69 | |
| XM_015772847.2 | XP_015628333.1 | 11 - 12 | 4252 | 69 | ||
| GenBank mRNA | AK108034.1 | - | 4 - 5 | 730 | 69 | |
More Information
Set Membership: |
|

LOC4334435
Genomic Map