Assay Details
Gene Symbol: LOC4335856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778019.2 | XP_015633505.1 | 3 - 4 | 586 | 63 | |
GenBank mRNA | AK073414.1 | - | 3 - 4 | 519 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
