Assay Details
Gene Symbol: LOC4335856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778019.2 | XP_015633505.1 | 5 - 6 | 768 | 83 | |
GenBank mRNA | AK073414.1 | - | 5 - 6 | 701 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
