Assay Details
Gene Symbol: LOC4335856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778019.2 | XP_015633505.1 | 6 - 7 | 856 | 74 | |
GenBank mRNA | AK073414.1 | - | 6 - 7 | 789 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
