Assay Details
| Gene Symbol: LOC4336254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780945.2 | XP_015636431.1 | 3 - 4 | 711 | 149 | |
| GenBank mRNA | AK120935.1 | - | 3 - 4 | 712 | 149 | |
More Information
Set Membership: |
|

LOC4336254
Genomic Map