Assay Details
| Gene Symbol: LOC4336810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781568.2 | XP_015637054.1 | 3 - 4 | 1971 | 81 | |
| GenBank mRNA | AK099941.1 | - | 3 - 4 | 1972 | 81 | |
More Information
Set Membership: |
|

LOC4336810
Genomic Map