Assay Details
| Gene Symbol: LOC4337469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781649.2 | XP_015637135.1 | 1 - 2 | 711 | 81 | |
| GenBank mRNA | AK121864.1 | - | 1 - 2 | 331 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781649.2 transcripts.
Set Membership: |
|

LOC4337469
Genomic Map