Assay Details
| Gene Symbol: LOC4337469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781649.2 | XP_015637135.1 | 6 - 7 | 3082 | 119 | |
| GenBank mRNA | AK121864.1 | - | 6 - 7 | 2702 | 119 | |
More Information
Set Membership: |
|

LOC4337469
Genomic Map