Assay Details
| Gene Symbol: LOC4337469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781649.2 | XP_015637135.1 | 7 - 8 | 3227 | 71 | |
| GenBank mRNA | AK121864.1 | - | 7 - 8 | 2847 | 71 | |
More Information
Set Membership: |
|

LOC4337469
Genomic Map