Assay Details
| Gene Symbol: LOC4339812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784747.2 | XP_015640233.1 | 2 - 3 | 374 | 65 | |
| GenBank mRNA | AB087745.1 | - | 2 - 3 | 337 | 65 | |
More Information
Set Membership: |
|

LOC4339812
Genomic Map