Assay Details
| Gene Symbol: LOC4339812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784747.2 | XP_015640233.1 | 5 - 6 | 831 | 69 | |
| GenBank mRNA | AB087745.1 | - | 5 - 6 | 794 | 69 | |
More Information
Set Membership: |
|

LOC4339812
Genomic Map