Assay Details
| Gene Symbol: LOC4324777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765396.2 | XP_015620882.1 | 10 - 11 | 1549 | 99 | |
| GenBank mRNA | AK101399.1 | - | 10 - 11 | 1164 | 99 | |
| AK102715.1 | - | 10 - 11 | 1386 | 99 | ||
More Information
Set Membership: |
|

LOC4324777
Genomic Map