Assay Details
| Gene Symbol: LOC4324777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765396.2 | XP_015620882.1 | 6 - 7 | 1142 | 115 | |
| GenBank mRNA | AK101399.1 | - | 6 - 7 | 757 | 115 | |
| AK102715.1 | - | 6 - 7 | 979 | 115 | ||
More Information
Set Membership: |
|

LOC4324777
Genomic Map