Assay Details
Gene Symbol: LOC4327742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758783.2 | XP_015614269.1 | 8 - 9 | 883 | 118 | |
GenBank mRNA | AK067201.1 | - | 2 - 3 | 154 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
