Assay Details
Gene Symbol: LOC4327742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758783.2 | XP_015614269.1 | 9 - 10 | 1003 | 116 | |
GenBank mRNA | AK067201.1 | - | 3 - 4 | 274 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
