Assay Details
Gene Symbol: LOC4327742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758783.2 | XP_015614269.1 | 10 - 11 | 1116 | 120 | |
GenBank mRNA | AK062088.1 | - | 1 - 2 | 121 | 120 | |
AK067201.1 | - | 4 - 5 | 387 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
