Assay Details
Gene Symbol: LOC4327742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758783.2 | XP_015614269.1 | 11 - 12 | 1237 | 77 | |
GenBank mRNA | AK062088.1 | - | 2 - 3 | 242 | 77 | |
AK067201.1 | - | 5 - 6 | 508 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
